Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nhr-25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029379.4 | NP_001024550.1 | 7 - 8 | 963 | 66 | |
NM_001029380.7 | NP_001024551.1 | 7 - 8 | 949 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |