Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: str-10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001322638.1 | NP_001309672.1 | 3 - 4 | 375 | 94 | |
NM_001322639.1 | NP_001309673.1 | 3 - 4 | 375 | 94 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |