Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cwp-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001373548.2 | NP_001359731.1 | 3 - 4 | 449 | 86 | |
NM_001373549.3 | NP_001359732.1 | 1 - 2 | 98 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |