Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gpa-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_077788.2 | NP_510189.1 | 5 - 6 | 658 | 143 | |
GenBank mRNA | AY008129.1 | - | 5 - 6 | 624 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Caenorhabditis elegans Probe spans exons |