Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nhr-32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_077800.7 | NP_510201.2 | 7 - 8 | 893 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |