Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srd-42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_077813.2 | NP_510214.2 | 3 - 4 | 620 | 82 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Recommended Assay Caenorhabditis elegans Probe spans exons |