Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: flr-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_077842.4 | NP_510243.1 | 9 - 10 | 1027 | 75 | |
GenBank mRNA | AB012617.1 | - | 9 - 10 | 933 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |