Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srd-50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001322645.3 | NP_001309446.1 | 4 - 5 | 720 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |