Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cdk-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_077855.7 | NP_510256.1 | 5 - 6 | 964 | 75 | |
GenBank mRNA | AF083878.1 | - | 4 - 5 | 777 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |