Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pab-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383665.1 | 5 - 6 | 1430 | 79 | ||
NM_077859.6 | NP_510260.1 | 4 - 5 | 1685 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |