Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: acr-12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001313441.4 | NP_001300370.1 | 9 - 10 | 1252 | 74 | |
NM_077861.6 | NP_510262.1 | 11 - 12 | 1715 | 74 | ||
GenBank mRNA | AF187013.1 | - | 11 - 12 | 1666 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |