Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cah-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_077863.8 | NP_510264.1 | 2 - 3 | 129 | 76 | |
NM_077864.5 | NP_510265.1 | 2 - 3 | 259 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |