Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: col-44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_077873.4 | NP_510274.1 | 1 - 2 | 194 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |