Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: twk-20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_077883.2 | NP_510284.1 | 10 - 11 | 912 | 107 | |
GenBank mRNA | AF083646.1 | - | 10 - 11 | 912 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |