Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: adm-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001368538.4 | NP_001355451.1 | 1 - 2 | 220 | 75 | |
NM_077890.6 | NP_510291.1 | 1 - 2 | 220 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |