Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pix-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270298.4 | NP_001257227.1 | 8 - 9 | 794 | 62 | |
NM_001270299.3 | NP_001257228.1 | 8 - 9 | 785 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |