Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: aakb-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001381137.1 | NP_001366890.1 | 3 - 4 | 494 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Caenorhabditis elegans Probe spans exons |