Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pqn-39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_077924.5 | NP_510325.2 | 7 - 8 | 2183 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |