Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C33G3.4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_077941.7 | NP_510342.1 | 13 - 14 | 2478 | 158 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |