Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sym-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_077960.7 | NP_510361.1 | 5 - 6 | 772 | 92 | |
GenBank mRNA | AY372076.1 | - | 3 - 4 | 563 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |