Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cyp-13B2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_077968.6 | NP_510369.2 | 3 - 4 | 456 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |