Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: irk-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001330918.4 | NP_001317893.1 | 10 - 11 | 1061 | 114 | |
NM_001330920.4 | NP_001317903.1 | 9 - 10 | 1031 | 114 | ||
NM_001330921.5 | NP_001317904.1 | 5 - 6 | 659 | 114 | ||
NM_001330922.4 | NP_001317905.1 | 3 - 4 | 308 | 114 | ||
NM_001381146.1 | NP_001368607.1 | 10 - 11 | 1076 | 114 | ||
NM_001381147.2 | NP_001366850.1 | 10 - 11 | 1120 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |