Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gfi-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078008.7 | NP_510409.1 | 13 - 14 | 1577 | 77 |
Gene Symbol: M163.8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001381148.1 | NP_001367098.1 | 13 - 14 | 1558 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |