Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sym-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078025.7 | NP_510426.1 | 1 - 2 | 258 | 73 | |
GenBank mRNA | AJ242473.1 | - | 1 - 2 | 162 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |