Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vit-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_076211.8 | NP_508612.1 | 1 - 2 | 866 | 139 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |