Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fbxb-43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_061588.3 | NP_493989.1 | 1 - 2 | 106 | 153 |
Gene Symbol: fbxb-42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_061589.4 | NP_493990.1 | 1 - 2 | 63 | 153 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |