Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srbc-27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001356782.1 | NP_001343600.1 | 4 - 5 | 633 | 157 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |