Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fbxa-94 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028698.4 | NP_001023869.1 | 2 - 3 | 184 | 75 | |
NM_001276970.3 | NP_001263899.1 | 1 - 2 | 58 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |