Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lea-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001269231.3 | NP_001256160.1 | 8 - 9 | 3762 | 92 | |
NM_001269232.3 | NP_001256161.1 | 7 - 8 | 3390 | 92 | ||
NM_001269234.3 | NP_001256163.1 | 7 - 8 | 3714 | 92 | ||
NM_001269235.3 | NP_001256164.1 | 6 - 7 | 3342 | 92 | ||
NM_001269240.3 | NP_001256169.1 | 6 - 7 | 3498 | 92 | ||
NM_001269241.4 | NP_001256170.1 | 5 - 6 | 3126 | 92 | ||
NM_001269244.3 | NP_001256173.1 | 1 - 2 | 1239 | 92 | ||
NM_001269245.3 | NP_001256174.1 | 1 - 2 | 1239 | 92 | ||
NM_001373085.2 | NP_001360513.1 | 8 | 3762 | 92 | ||
NM_001373086.2 | NP_001360514.1 | 7 | 3390 | 92 | ||
NM_001373087.2 | NP_001360515.1 | 7 | 3714 | 92 | ||
NM_001373088.2 | NP_001360516.1 | 6 - 7 | 3342 | 92 | ||
NM_001373089.2 | NP_001360517.1 | 6 | 3498 | 92 | ||
NM_001373090.2 | NP_001360518.1 | 5 | 3126 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Amplicon spans exons and probe does not span exons |