Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bus-17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029832.5 | NP_001025003.2 | 6 - 7 | 803 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |