Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gpa-18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001306789.3 | NP_001293718.1 | 6 - 7 | 915 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |