Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srt-33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_074134.2 | NP_506535.2 | 3 - 4 | 935 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |