Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: swp-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025951.3 | NP_001021122.1 | 1 - 2 | 49 | 109 | |
NM_001379724.2 | NP_001367326.1 | 1 - 2 | 52 | 109 | ||
NM_001382877.1 | 1 - 2 | 54 | 109 | |||
NM_001382878.1 | 1 - 2 | 58 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |