Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fut-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027139.2 | NP_001022310.1 | 2 - 3 | 322 | 118 | |
GenBank mRNA | AJ745073.1 | - | 2 - 3 | 238 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |