Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srh-42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027301.3 | NP_001022472.1 | 2 - 3 | 927 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |