Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: grl-25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027854.5 | NP_001023025.1 | 5 - 6 | 2145 | 100 | |
NR_101843.1 | 5 - 6 | 2585 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |