Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srsx-16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268344.2 | NP_001255273.1 | 2 - 3 | 275 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |