Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srv-28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028212.1 | NP_001023383.1 | 4 - 5 | 745 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Detect Genomic DNA Caenorhabditis elegans Probe spans exons |