Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: F26A1.19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001346611.3 | NP_001333555.1 | 2 - 3 | 205 | 95 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Caenorhabditis elegans Probe spans exons |