Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lgc-29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038424.4 | NP_001033513.2 | 4 - 5 | 472 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |