Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047215.5 | NP_001040680.1 | 5 - 6 | 859 | 66 | |
NM_001047216.3 | NP_001040681.1 | 1 - 2 | 30 | 66 | ||
NM_001129040.5 | NP_001122512.1 | 5 - 6 | 867 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Caenorhabditis elegans Probe spans exons |