Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fbxa-55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047409.4 | NP_001040874.1 | 2 - 3 | 765 | 153 | |
NM_001330938.4 | NP_001317783.1 | 3 | 676 | 153 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |