Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRPC7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022425051.1 | XP_022280759.1 | 5 - 6 | 1527 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Canis familiaris Probe spans exons |