Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC8B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005621953.3 | XP_005622010.1 | 4 - 5 | 2389 | 88 | |
XM_022420527.1 | XP_022276235.1 | 5 - 6 | 2533 | 88 | ||
XM_022420528.1 | XP_022276236.1 | 4 - 5 | 2389 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Canis familiaris Probe spans exons |