Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUP58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003433299.4 | XP_003433347.1 | 1587 | 75 | ||
XM_005635541.3 | XP_005635598.1 | 1625 | 75 | |||
XM_534528.6 | XP_534528.2 | 1624 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Canis familiaris |