Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KMT5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005636119.3 | XP_005636176.1 | 748 | 75 | ||
XM_005636120.3 | XP_005636177.1 | 677 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Canis familiaris |