Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STEAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005628485.3 | XP_005628542.1 | 3 - 4 | 690 | 91 | |
XM_014119219.2 | XP_013974694.1 | 3 - 4 | 705 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Canis familiaris Inventoried Probe spans exons |