Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FCER1G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003171.2 | NP_001003171.2 | 4 - 5 | 194 | 89 | |
XM_014111133.2 | XP_013966608.1 | 5 - 6 | 586 | 89 | ||
GenBank mRNA | AB017553.1 | - | 4 - 5 | 194 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Canis familiaris Probe spans exons |