Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTGDS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003131.1 | NP_001003131.1 | 2 - 3 | 308 | 64 | |
GenBank mRNA | AB026988.1 | - | 2 - 3 | 308 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Canis familiaris Probe spans exons |