Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MT2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003149.1 | NP_001003149.1 | 1 - 2 | 86 | 108 | |
GenBank mRNA | AB028042.1 | - | 1 - 2 | 86 | 108 | |
AJ388530.1 | - | 1 - 2 | 71 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Canis familiaris Inventoried Probe spans exons |